Product Details

SNP ID
rs16832823
Assay Type
Functionally tested
NCBI dbSNP Submissions
33
Location
Chr.1:161321533 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
TGGGCCTAGCCTGGTGTGAGGAATG[C/T]GTGCCCTGCAACTTCAGAGTCACAA
Phenotype
MIM: 602413
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
SDHC PubMed Links

Gene Details

Gene
SDHC
Gene Name
succinate dehydrogenase complex subunit C
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001035511.1 Intron NP_001030588.1
NM_001035512.1 Intron NP_001030589.1
NM_001035513.1 Intron NP_001030590.1
NM_001278172.1 Intron NP_001265101.1
NM_003001.3 Intron NP_002992.1

View Full Product Details