Product Details

SNP ID
rs17666900
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.5:179113014 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
CATTACTTTCGCAGGTGGAGGACAG[A/C]TGCACGGCTCAGACTCGGTCGAGAC
Phenotype
MIM: 604539
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
ADAMTS2 PubMed Links

Gene Details

Gene
ADAMTS2
Gene Name
ADAM metallopeptidase with thrombospondin type 1 motif 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_014244.4 4591 UTR 3 NP_055059.2
NM_021599.3 4591 Intron NP_067610.1
XM_017010063.1 4591 UTR 3 XP_016865552.1

View Full Product Details