Transcript Accession | SNP Location | SNP Type | Codon Change | Amino Acid Change | Protein ID |
---|---|---|---|---|---|
NM_000135.2 | 4799 | UTR 3 | NP_000126.2 | ||
NM_001018112.1 | 4799 | Intron | NP_001018122.1 | ||
NM_001286167.1 | 4799 | UTR 3 | NP_001273096.1 | ||
XM_005256294.4 | 4799 | Intron | XP_005256351.1 | ||
XM_011522945.2 | 4799 | Intron | XP_011521247.1 | ||
XM_011522946.2 | 4799 | Intron | XP_011521248.1 | ||
XM_011522947.2 | 4799 | Intron | XP_011521249.1 | ||
XM_011522948.2 | 4799 | Intron | XP_011521250.1 | ||
XM_017023044.1 | 4799 | Intron | XP_016878533.1 | ||
XM_017023045.1 | 4799 | Intron | XP_016878534.1 | ||
XM_017023046.1 | 4799 | Intron | XP_016878535.1 |
Transcript Accession | SNP Location | SNP Type | Codon Change | Amino Acid Change | Protein ID |
---|---|---|---|---|---|
NM_001113525.1 | 4799 | Silent Mutation | GAC,GAG | D605E | NP_001106997.1 |
NM_152287.3 | 4799 | Silent Mutation | GAC,GAG | D530E | NP_689500.2 |
XM_005256324.3 | 4799 | Silent Mutation | GAC,GAG | D588E | XP_005256381.1 |
XM_005256328.3 | 4799 | Silent Mutation | GAC,GAG | D391E | XP_005256385.1 |
XM_017023889.1 | 4799 | Intron | XP_016879378.1 | ||
XM_017023890.1 | 4799 | Silent Mutation | GAC,GAG | D391E | XP_016879379.1 |