Product Details

SNP ID
rs6509867
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:54354326 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
CTGTAGGGTCGGCTCTCTGCCTTCA[A/C]CCAGCCACACCTCCCTCATTCATTC
Phenotype
MIM: 602992
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
LAIR1 PubMed Links

Gene Details

Gene
LAIR1
Gene Name
leukocyte associated immunoglobulin like receptor 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001289023.2 2116 UTR 3 NP_001275952.2
NM_001289025.2 2116 UTR 3 NP_001275954.2
NM_001289026.2 2116 UTR 3 NP_001275955.2
NM_001289027.2 2116 UTR 3 NP_001275956.2
NM_002287.5 2116 UTR 3 NP_002278.2
NM_021706.4 2116 UTR 3 NP_068352.2
XM_017026803.1 2116 Intron XP_016882292.1

View Full Product Details