Transcript Accession | SNP Location | SNP Type | Codon Change | Amino Acid Change | Protein ID |
---|---|---|---|---|---|
NM_000502.5 | 2241 | Intron | NP_000493.1 |
Transcript Accession | SNP Location | SNP Type | Codon Change | Amino Acid Change | Protein ID |
---|---|---|---|---|---|
NM_001165927.1 | 2241 | UTR 3 | NP_001159399.1 | ||
NM_001321268.1 | 2241 | UTR 3 | NP_001308197.1 | ||
NM_001321269.1 | 2241 | UTR 3 | NP_001308198.1 | ||
NM_017777.3 | 2241 | UTR 3 | NP_060247.2 | ||
XM_005257485.3 | 2241 | UTR 3 | XP_005257542.1 | ||
XM_006721965.2 | 2241 | UTR 3 | XP_006722028.1 | ||
XM_011524957.2 | 2241 | UTR 3 | XP_011523259.1 | ||
XM_011524958.2 | 2241 | UTR 3 | XP_011523260.1 | ||
XM_011524959.2 | 2241 | UTR 3 | XP_011523261.1 | ||
XM_011524960.2 | 2241 | Intron | XP_011523262.1 | ||
XM_017024803.1 | 2241 | UTR 3 | XP_016880292.1 | ||
XM_017024804.1 | 2241 | Intron | XP_016880293.1 | ||
XM_017024805.1 | 2241 | UTR 3 | XP_016880294.1 |