Product Details

SNP ID
rs17118576
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.12:56417212 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
ACTATATTCCAACCCTATCTCAAAT[A/G]CAGGAATGGGCCTCTATCTGGTATC
Phenotype
MIM: 603887
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
TIMELESS PubMed Links

Gene Details

Gene
TIMELESS
Gene Name
timeless circadian clock
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
XM_011538939.2 4274 UTR 3 XP_011537241.1

View Full Product Details