Product Details

SNP ID
rs17853366
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.2:73075688 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CTGATGAGCTCATCGTGGGTCAGGT[G/T]GTAGTACTTGGCCGACTGGTCCAGC
Phenotype
MIM: 605536 MIM: 615572
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
RAB11FIP5 PubMed Links

Gene Details

Gene
RAB11FIP5
Gene Name
RAB11 family interacting protein 5
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_015470.2 3990 Missense Mutation AAC,CAC N599H NP_056285.1
XM_005264251.3 3990 Missense Mutation AAC,CAC N1164H XP_005264308.1
XM_005264252.3 3990 Missense Mutation AAC,CAC N1270H XP_005264309.1
XM_005264253.4 3990 Intron XP_005264310.1
XM_006711985.3 3990 Missense Mutation AAC,CAC N1195H XP_006712048.1
XM_011532753.2 3990 Missense Mutation AAC,CAC N1064H XP_011531055.1
XM_011532754.2 3990 Intron XP_011531056.1
XM_011532755.2 3990 Intron XP_011531057.1
XM_011532756.2 3990 Intron XP_011531058.1
XM_017003788.1 3990 Missense Mutation AAC,CAC N1019H XP_016859277.1
Gene
SFXN5
Gene Name
sideroflexin 5
There are no transcripts associated with this gene.

View Full Product Details