Product Details

SNP ID
rs6728610
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.2:37644080 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
CATGTGGCCCAAGAAAATTCTTCCA[A/T]TGCGGTCCAGGGAAGCTAAAATATT
Phenotype
MIM: 606133
Polymorphism
A/T, Transversion substitution
Allele Nomenclature
Literature Links
CDC42EP3 PubMed Links

Gene Details

Gene
CDC42EP3
Gene Name
CDC42 effector protein 3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001270436.1 3517 UTR 3 NP_001257365.1
NM_001270437.1 3517 UTR 3 NP_001257366.1
NM_001270438.1 3517 UTR 3 NP_001257367.1
NM_006449.4 3517 UTR 3 NP_006440.2
XM_011532482.2 3517 UTR 3 XP_011530784.1

View Full Product Details