Product Details

SNP ID
rs17018467
Assay Type
Functionally tested
NCBI dbSNP Submissions
13
Location
Chr.1:107147599 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
CAATGCGGATTTTCACTCCAGCAAA[C/G]AAAAGCTAAGAAAAGTACAAGGAAT
Phenotype
MIM: 608818
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
NTNG1 PubMed Links

Gene Details

Gene
NTNG1
Gene Name
netrin G1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001113226.2 Intron NP_001106697.1
NM_001113228.2 Intron NP_001106699.1
NM_001312688.1 Intron NP_001299617.1
NM_014917.3 Intron NP_055732.2
XM_006710455.2 Intron XP_006710518.1
XM_006710456.3 Intron XP_006710519.1
XM_011541021.1 Intron XP_011539323.1
XM_011541024.2 Intron XP_011539326.1
XM_011541025.1 Intron XP_011539327.1
XM_017000680.1 Intron XP_016856169.1
XM_017000681.1 Intron XP_016856170.1
XM_017000682.1 Intron XP_016856171.1
XM_017000683.1 Intron XP_016856172.1
XM_017000684.1 Intron XP_016856173.1
XM_017000685.1 Intron XP_016856174.1
XM_017000686.1 Intron XP_016856175.1

View Full Product Details