Product Details

SNP ID
rs16973887
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:37563959 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
AAAACTTTTGCTTTTGGAATTTGTT[A/G]TAGAAAAATAATCTTTCACTTTCTT
Phenotype
MIM: 613903
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
ZNF540 PubMed Links

Gene Details

Gene
ZNF540
Gene Name
zinc finger protein 540
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001172225.2 Intron NP_001165696.1
NM_001172226.2 Intron NP_001165697.1
NM_152606.4 Intron NP_689819.1
Gene
ZNF571
Gene Name
zinc finger protein 571
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001290314.1 Intron NP_001277243.1
NM_001321272.1 Intron NP_001308201.1
NM_016536.4 Intron NP_057620.3
XM_011527024.2 Intron XP_011525326.1
XM_017026855.1 Intron XP_016882344.1
XM_017026856.1 Intron XP_016882345.1
XM_017026857.1 Intron XP_016882346.1
XM_017026858.1 Intron XP_016882347.1
XM_017026859.1 Intron XP_016882348.1
XM_017026860.1 Intron XP_016882349.1
Gene
ZNF571-AS1
Gene Name
ZNF571 antisense RNA 1
There are no transcripts associated with this gene.

View Full Product Details