Product Details

SNP ID
rs17468865
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.9:36039884 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CTGTAGAAAGAGCTAGGATTGGTGA[C/T]AGACCAGATATGACAGTTTATTCTG
Phenotype
MIM: 605227
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
RECK PubMed Links

Gene Details

Gene
RECK
Gene Name
reversion inducing cysteine rich protein with kazal motifs
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001316345.1 Intron NP_001303274.1
NM_001316346.1 Intron NP_001303275.1
NM_001316347.1 Intron NP_001303276.1
NM_001316348.1 Intron NP_001303277.1
NM_021111.2 Intron NP_066934.1
XM_017015207.1 Intron XP_016870696.1
XM_017015208.1 Intron XP_016870697.1
XM_017015209.1 Intron XP_016870698.1
XM_017015210.1 Intron XP_016870699.1
XM_017015211.1 Intron XP_016870700.1

View Full Product Details