Product Details

SNP ID
rs17855037
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.9:36037036 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CGGGCCTCTCTGCGAGGTGCGCTGC[C/T]CCTTCTGCTGGCCGTGGCGGGGGTC
Phenotype
MIM: 605227
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
RECK PubMed Links

Gene Details

Gene
RECK
Gene Name
reversion inducing cysteine rich protein with kazal motifs
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001316345.1 126 UTR 5 NP_001303274.1
NM_001316346.1 126 Missense Mutation CCC,CTC P13L NP_001303275.1
NM_001316347.1 126 Missense Mutation CCC,CTC P13L NP_001303276.1
NM_001316348.1 126 Missense Mutation CCC,CTC P13L NP_001303277.1
NM_021111.2 126 Missense Mutation CCC,CTC P13L NP_066934.1
XM_017015207.1 126 UTR 5 XP_016870696.1
XM_017015208.1 126 UTR 5 XP_016870697.1
XM_017015209.1 126 UTR 5 XP_016870698.1
XM_017015210.1 126 Intron XP_016870699.1
XM_017015211.1 126 Missense Mutation CCC,CTC P13L XP_016870700.1

View Full Product Details