Product Details

SNP ID
rs7864610
Assay Type
Functionally Tested
NCBI dbSNP Submissions
NA
Location
Chr.9:4802448 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
ATATGTCCATTTGGGGATAATTGTC[A/G]TCTTTATTATGTTGAGGTTTTAATC
Phenotype
MIM: 611405
Polymorphism
A/G, Transition Substitution
Allele Nomenclature
Literature Links
RCL1 PubMed Links
Additional Information
For this assay, SNP(s) [rs76234352] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.

Gene Details

Gene
RCL1
Gene Name
RNA terminal phosphate cyclase like 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001286699.1 Intron NP_001273628.1
NM_001286700.1 Intron NP_001273629.1
NM_001286701.1 Intron NP_001273630.1
NM_005772.4 Intron NP_005763.3
XM_006716715.3 Intron XP_006716778.1
XM_011517673.2 Intron XP_011515975.1
XM_017014176.1 Intron XP_016869665.1

View Full Product Details