Product Details

SNP ID
rs17001846
Assay Type
Functionally Tested
NCBI dbSNP Submissions
NA
Location
Chr.22:40348049 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
GAGTATGTGTTCTTTGTGTTGGCCT[A/G]AAATGCATACAATTTTGGCTTTGGA
Phenotype
MIM: 608222
Polymorphism
A/G, Transition Substitution
Allele Nomenclature
Literature Links
ADSL PubMed Links

Gene Details

Gene
ADSL
Gene Name
adenylosuccinate lyase
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_000026.3 Intron NP_000017.1
NM_001123378.2 Intron NP_001116850.1
NM_001317923.1 Intron NP_001304852.1
XM_011529977.2 Intron XP_011528279.1
XM_011529979.1 Intron XP_011528281.1
XM_011529980.2 Intron XP_011528282.1
XM_017028636.1 Intron XP_016884125.1
XM_017028637.1 Intron XP_016884126.1
XM_017028638.1 Intron XP_016884127.1
XM_017028639.1 Intron XP_016884128.1
XM_017028640.1 Intron XP_016884129.1

View Full Product Details