Product Details

SNP ID
rs17074611
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.3:42582424 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GCACAACCTACTGCCAGCCTGGATG[A/G]GGCGCCAAAGTCCGACCCAGGGGGA
Phenotype
MIM: 604028 MIM: 606473
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
SEC22C PubMed Links

Gene Details

Gene
SEC22C
Gene Name
SEC22 homolog C, vesicle trafficking protein
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001201572.1 Intron NP_001188501.1
NM_001201584.1 Intron NP_001188513.1
NM_004206.3 Intron NP_004197.1
NM_032970.3 Intron NP_116752.1
XM_005265533.1 Intron XP_005265590.1
XM_005265534.1 Intron XP_005265591.1
XM_005265535.1 Intron XP_005265592.1
XM_011534212.1 Intron XP_011532514.1
XM_017007438.1 Intron XP_016862927.1
XM_017007439.1 Intron XP_016862928.1
Gene
SS18L2
Gene Name
SS18 like 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_016305.3 Intron NP_057389.1
XM_017006548.1 Intron XP_016862037.1
XM_017006549.1 Intron XP_016862038.1

View Full Product Details