Product Details

SNP ID
rs17101758
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.14:24176755 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
CCTTGACTTCAGAGTCCATGCTTTT[C/T]CTTAACTGCATGGCTGTGGGAATTT
Phenotype
MIM: 608193
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
IPO4 PubMed Links

Gene Details

Gene
IPO4
Gene Name
importin 4
There are no transcripts associated with this gene.

Gene
REC8
Gene Name
REC8 meiotic recombination protein
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001048205.1 Intron NP_001041670.1
NM_005132.2 Intron NP_005123.2
XM_011537421.1 Intron XP_011535723.1
XM_017021841.1 Intron XP_016877330.1

View Full Product Details