Product Details

SNP ID
rs17490906
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:112256803 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
CCTGACATCTGTTCTAGATCTGTGC[G/T]TCAGGTGCAGTAGAAGGTGAGCAGA
Phenotype
MIM: 611904
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
LOC100132686 PubMed Links

Gene Details

Gene
LOC100132686
Gene Name
uncharacterized LOC100132686
There are no transcripts associated with this gene.

Gene
PLET1
Gene Name
placenta expressed transcript 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001145024.1 Intron NP_001138496.1

View Full Product Details