Product Details

SNP ID
rs17852004
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.2:231712450 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GCCTTTGATTCTCTCCAGGTGAGGA[A/G]GAGGATGGAGATGAAGATGAGGAAG
Phenotype
MIM: 188390
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
MIR1244-1 PubMed Links

Gene Details

Gene
MIR1244-1
Gene Name
microRNA 1244-1
There are no transcripts associated with this gene.

Gene
PTMA
Gene Name
prothymosin, alpha
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001099285.1 1030 Intron NP_001092755.1
NM_002823.4 1030 Intron NP_002814.3
XM_011511518.1 1030 Silent Mutation GAA,GAG E226E XP_011509820.1
XM_011511519.1 1030 Silent Mutation GAA,GAG E225E XP_011509821.1
XM_011511520.1 1030 Silent Mutation GAA,GAG E225E XP_011509822.1
XM_011511521.1 1030 Silent Mutation GAA,GAG E224E XP_011509823.1
XM_011511522.2 1030 Missense Mutation AAG,AGG K194R XP_011509824.1

View Full Product Details