Product Details

SNP ID
rs17092209
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.20:35004918 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
GATGGATCCATGACGCTGCATTAAG[A/G]AAAACAACGCAAAAGTTCCACTCCT
Phenotype
MIM: 609928 MIM: 608430
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
MYH7B PubMed Links

Gene Details

Gene
MYH7B
Gene Name
myosin heavy chain 7B
There are no transcripts associated with this gene.

Gene
TRPC4AP
Gene Name
transient receptor potential cation channel subfamily C member 4 associated protein
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_015638.2 Intron NP_056453.1
NM_199368.1 Intron NP_955400.1
XM_011528772.2 Intron XP_011527074.1
XM_011528774.1 Intron XP_011527076.1
XM_017027799.1 Intron XP_016883288.1
XM_017027800.1 Intron XP_016883289.1
XM_017027801.1 Intron XP_016883290.1

View Full Product Details