Product Details

SNP ID
rs17092750
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.20:35527132 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
ACACGCTAGCTGATGGATGACTTCG[A/T]TTTCTGCTCCTGCACTAGACGTGGG
Phenotype
MIM: 609689
Polymorphism
A/T, Transversion substitution
Allele Nomenclature
Literature Links
C20orf173 PubMed Links

Gene Details

Gene
C20orf173
Gene Name
chromosome 20 open reading frame 173
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001145350.1 898 UTR 3 NP_001138822.1
XM_006723710.2 898 UTR 3 XP_006723773.1
XM_011528577.1 898 Intron XP_011526879.1
Gene
CEP250
Gene Name
centrosomal protein 250
There are no transcripts associated with this gene.

View Full Product Details