Product Details

SNP ID
rs35400877
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.16:1494300 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGCAGGATGGAGCCAGCACCCTCA[CG/GA]GGTTCGACTCGCCGTCCGGGAAGC
Phenotype
MIM: 613442 MIM: 611140
Polymorphism
CG/GA, Multiple nucleotide polymorphism
Allele Nomenclature
Literature Links
PTX4 PubMed Links

Gene Details

Gene
PTX4
Gene Name
pentraxin 4
There are no transcripts associated with this gene.

Gene
TELO2
Gene Name
telomere maintenance 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_016111.3 369 Silent Mutation GAA,GAG E7E NP_057195.2
XM_011522773.2 369 Missense Mutation CGA,GAG R7E XP_011521075.1
XM_011522774.2 369 Missense Mutation CGA,GAG R7E XP_011521076.1
XM_011522775.2 369 Missense Mutation CGA,GAG R7E XP_011521077.1
XM_011522776.2 369 Silent Mutation GAA,GAG E7E XP_011521078.1
XM_011522777.2 369 Missense Mutation CGA,GAG R7E XP_011521079.1
XM_011522778.2 369 Missense Mutation CGA,GAG R7E XP_011521080.1
XM_017023914.1 369 Missense Mutation CGA,GAG R7E XP_016879403.1

View Full Product Details