Product Details
- SNP ID
-
rs17019726
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
31
- Location
-
Chr.1:212795786 on Build GRCh38
- Set Membership
-
HapMap
- Context Sequence [VIC/FAM]
- GCCTTTCTCGACTTTACCTCATTCA[A/G]TGTTTCACAATAGCCTTCTGAGGAA
- Phenotype
-
MIM: 609174
- Polymorphism
- A/G, Transition Substitution
- Allele Nomenclature
-
- Literature Links
-
NSL1
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs138702367] are located under a probe and SNP(s) [rs75366521] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- NSL1
- Gene Name
- NSL1, MIS12 kinetochore complex component
- Gene
- TATDN3
- Gene Name
- TatD DNase domain containing 3
View Full Product Details