Product Details
- SNP ID
-
rs121965073
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.15:80153101 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- GAGGATTCCGACTTCCCCATCCACA[A/T]CCTGCCCTACGGCGTCTTCTCGACC
- Phenotype
-
MIM: 613871
- Polymorphism
- A/T, Transversion substitution
- Allele Nomenclature
-
- Literature Links
-
FAH
PubMed Links
Gene Details
- Gene
- FAH
- Gene Name
- fumarylacetoacetate hydrolase
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_000137.2 |
211 |
Missense Mutation |
AAC,ATC |
N16I |
NP_000128.1 |
View Full Product Details