Product Details

SNP ID
rs17344661
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.13:21143383 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
TACACTAGAGCTAATCCCTTGAAGA[C/T]TGGGACCTTACTTGTCTTATTTGGA
Phenotype
MIM: 602949
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
SAP18 PubMed Links

Gene Details

Gene
SAP18
Gene Name
Sin3A associated protein 18
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_005870.4 Intron NP_005861.2
XM_011534868.2 Intron XP_011533170.1

View Full Product Details