Product Details

SNP ID
rs11545380
Assay Type
Functionally tested
NCBI dbSNP Submissions
1
Location
Chr.1:43305283 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ACACACTGTGAACAAAGGTGACACC[A/G]CTGTACTTTCTGCACGTGTGCACAA
Phenotype
MIM: 600222
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
TIE1 PubMed Links

Gene Details

Gene
TIE1
Gene Name
tyrosine kinase with immunoglobulin like and EGF like domains 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001253357.1 518 Missense Mutation ACT,GCT T97A NP_001240286.1
NM_005424.4 518 Missense Mutation ACT,GCT T142A NP_005415.1
XM_005271163.1 518 Missense Mutation ACT,GCT T142A XP_005271220.1
XM_006710869.1 518 UTR 5 XP_006710932.1
XM_017002207.1 518 Missense Mutation ACT,GCT T142A XP_016857696.1

View Full Product Details