Product Details

SNP ID
hCV42656416
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.2:216415322 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CCCCTTCGGGGCAGAACATTTCTTA[C/G]ATCCATTCTAGCTCAGAGAGTGTAA
Phenotype
MIM: 606622
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
SMARCAL1 PubMed Links

Gene Details

Gene
SMARCAL1
Gene Name
SWI/SNF related, matrix associated, actin dependent regulator of chromatin, subfamily a like 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001127207.1 762 Nonsense Mutation TAC,TAG Y206* NP_001120679.1
NM_014140.3 762 Nonsense Mutation TAC,TAG Y206* NP_054859.2

View Full Product Details