Product Details

SNP ID
rs6780162
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.3:114215179 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
TTTAGTGAGTTAAAAAAAAAACCAG[C/T]AAACCAACACAAGACTCATAAGTAC
Phenotype
MIM: 194553
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
ZNF80 PubMed Links

Gene Details

Gene
ZNF80
Gene Name
zinc finger protein 80
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_007136.3 Intron NP_009067.2
XM_017007132.1 Intron XP_016862621.1
XM_017007133.1 Intron XP_016862622.1
XM_017007134.1 Intron XP_016862623.1
XM_017007135.1 Intron XP_016862624.1
XM_017007136.1 Intron XP_016862625.1

View Full Product Details