Product Details

SNP ID
rs11556244
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.3:139347021 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ACTGAAGCCACCAACCTATAAGCTA[A/G]TGACTCAGGCACAGTTGGAAGAGGT
Phenotype
MIM: 606990 MIM: 605810
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
COPB2 PubMed Links

Gene Details

Gene
COPB2
Gene Name
coatomer protein complex subunit beta 2
There are no transcripts associated with this gene.

Gene
MRPS22
Gene Name
mitochondrial ribosomal protein S22
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_020191.2 273 Missense Mutation ATG,GTG M106V NP_064576.1
XM_005247640.2 273 Missense Mutation ATG,GTG M105V XP_005247697.1
XM_006713703.3 273 Missense Mutation ATG,GTG M106V XP_006713766.1
XM_011512995.2 273 Missense Mutation ATG,GTG M65V XP_011511297.1
XM_011512996.2 273 Missense Mutation ATG,GTG M64V XP_011511298.1

View Full Product Details