Product Details

SNP ID
rs7631399
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.3:101523626 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GATGAACTGGGAATTCAGTGTCATC[A/G]TTAATATTTTACAGTGTTAGTTAAG
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
FAM172BP PubMed Links

Gene Details

Gene
FAM172BP
Gene Name
family with sequence similarity 172, member B pseudogene
There are no transcripts associated with this gene.

View Full Product Details