Product Details

SNP ID
rs12233490
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.3:185512302 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CTCCCTCTCAAGATGAACCTTAAAA[A/G]AGGCAAATCCAGAGTCTGATCCTGT
Phenotype
MIM: 607365
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
LIPH PubMed Links

Gene Details

Gene
LIPH
Gene Name
lipase H
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_139248.2 Intron NP_640341.1
XM_006713529.3 Intron XP_006713592.1
XM_011512530.2 Intron XP_011510832.1
XM_011512531.2 Intron XP_011510833.1
XM_017005852.1 Intron XP_016861341.1

View Full Product Details