Product Details

SNP ID
rs12639755
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.4:8278313 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
GGATGTGACACCGGACACCTAGCCT[C/T]GGGCCTTGATTGCTTTGCTCTGTGT
Phenotype
MIM: 608785
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
HTRA3 PubMed Links

Gene Details

Gene
HTRA3
Gene Name
HtrA serine peptidase 3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001297559.1 Intron NP_001284488.1
NM_053044.4 Intron NP_444272.1
XM_011513596.2 Intron XP_011511898.1

View Full Product Details