Product Details

SNP ID
rs7715390
Assay Type
Functionally Tested
NCBI dbSNP Submissions
NA
Location
Chr.5:83055897 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ATCTAATTAATACTATCTACAACAG[G/T]TAATTTTTAGCTAAGATTTAAGTTC
Phenotype
MIM: 611329
Polymorphism
G/T, Transversion Substitution
Allele Nomenclature
Literature Links
SCARNA18 PubMed Links
Additional Information
For this assay, SNP(s) [rs140928522] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.

Gene Details

Gene
SCARNA18
Gene Name
small Cajal body-specific RNA 18
There are no transcripts associated with this gene.

Gene
TMEM167A
Gene Name
transmembrane protein 167A
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_174909.4 Intron NP_777569.1

View Full Product Details