Product Details

SNP ID
rs4865934
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.5:55313130 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
GATTTTTATAAATGTATCATTTCTC[A/G]GTTTTGTTGAAAATCCTCATTCCTA
Phenotype
MIM: 612720
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
DHX29 PubMed Links

Gene Details

Gene
DHX29
Gene Name
DEAH-box helicase 29
There are no transcripts associated with this gene.

Gene
SKIV2L2
Gene Name
Ski2 like RNA helicase 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_015360.4 Intron NP_056175.3

View Full Product Details