Product Details
- SNP ID
-
rs11548798
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.7:25124043 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- ATGGAGATTTGGCCCAGTCTTGTGC[A/T]TGCCTCCCTTTTCAACGGTGTGGCA
- Phenotype
-
MIM: 123970
- Polymorphism
- A/T, Transversion substitution
- Allele Nomenclature
-
- Literature Links
-
CYCS
PubMed Links
Gene Details
- Gene
- CYCS
- Gene Name
- cytochrome c, somatic
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_018947.5 |
247 |
Missense Mutation |
AAG,ATG |
K26M |
NP_061820.1 |
View Full Product Details