Product Details

SNP ID
rs11547167
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.7:76302835 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TCGGGCTGCCCCGGCTGCCGGAGGA[G/T]TGGTCGCAGTGGTTAGGCGGCAGCA
Phenotype
MIM: 602195
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
HSPB1 PubMed Links

Gene Details

Gene
HSPB1
Gene Name
heat shock protein family B (small) member 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001540.3 278 Missense Mutation GAG,GAT E41D NP_001531.1

View Full Product Details