Product Details

SNP ID
rs11547071
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.9:128163485 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TCCATGCTGGACCAGATCAACTCCT[C/G]TCTGGACCACCTGGAGGAGAAGAAT
Phenotype
MIM: 611420
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
C9orf16 PubMed Links

Gene Details

Gene
C9orf16
Gene Name
chromosome 9 open reading frame 16
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_024112.3 270 Missense Mutation TCT,TGT S41C NP_077017.1
XM_011519004.1 270 Missense Mutation TCT,TGT S68C XP_011517306.1
Gene
CIZ1
Gene Name
CDKN1A interacting zinc finger protein 1
There are no transcripts associated with this gene.

View Full Product Details