Product Details

SNP ID
rs7303801
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.12:8226412 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
CACGGTAGGTCTGTCAGCAGAGAAC[C/G]CTGACCACACACTCATGTGTTTTCT
Phenotype
MIM: 613041
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
FAM86FP PubMed Links

Gene Details

Gene
FAM86FP
Gene Name
family with sequence similarity 86, member A pseudogene
There are no transcripts associated with this gene.

Gene
FAM90A1
Gene Name
family with sequence similarity 90 member A1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001319982.1 Intron NP_001306911.1
NM_018088.3 Intron NP_060558.3
XM_006719097.3 Intron XP_006719160.1
XM_011520719.1 Intron XP_011519021.1
XM_017019547.1 Intron XP_016875036.1
Gene
LOC101927905
Gene Name
uncharacterized LOC101927905
There are no transcripts associated with this gene.

View Full Product Details