Product Details
- SNP ID
-
hCV43523525
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.15:40333267 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- CCAGCATGGGCTTCACCTGAAGCCT[C/T]TTAGAAATACGAATTCTCGGCCAGG
- Phenotype
-
- Polymorphism
- C/T, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
C15orf52
PubMed Links
Gene Details
- Gene
- C15orf52
- Gene Name
- chromosome 15 open reading frame 52
There are no transcripts associated with this gene.
- Gene
- INAFM2
- Gene Name
- InaF motif containing 2
There are no transcripts associated with this gene.
View Full Product Details