Product Details
- SNP ID
-
rs12914738
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.15:75600203 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- GGAAAATGTGCTTGTTGAGCCTAGA[A/G]AAGGTAGCTGCAGGTTCTGAGGCTG
- Phenotype
-
MIM: 607902
- Polymorphism
- A/G, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
SNUPN
PubMed Links
Gene Details
- Gene
- SNUPN
- Gene Name
- snurportin 1
View Full Product Details