Product Details

SNP ID
rs1803555
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.15:76225862 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AGGCCAGCTTACCTTAAATAAATCT[G/T]CAACTATTCCATAATCTGCCACTTG
Phenotype
MIM: 608053
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
ETFA PubMed Links

Gene Details

Gene
ETFA
Gene Name
electron transfer flavoprotein alpha subunit
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_000126.3 1031 Missense Mutation GAA,GCA E317A NP_000117.1
NM_001127716.1 1031 Missense Mutation GAA,GCA E268A NP_001121188.1

View Full Product Details