Product Details

SNP ID
rs11862413
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.16:74675308 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
ATGGGGAATTTCTGGTCTACTGGAA[A/G]GGGACCCAACCTCACCACTGGCTGA
Phenotype
MIM: 615153 MIM: 614151
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
MLKL PubMed Links

Gene Details

Gene
MLKL
Gene Name
mixed lineage kinase domain-like
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001142497.2 Intron NP_001135969.1
NM_152649.3 Intron NP_689862.1
XM_005255834.3 Intron XP_005255891.1
XM_011522936.2 Intron XP_011521238.1
Gene
RFWD3
Gene Name
ring finger and WD repeat domain 3
There are no transcripts associated with this gene.

View Full Product Details