Product Details

SNP ID
rs11547593
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.16:84295005 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AACCGGCGTGGGGCCGGGCAGCTGC[A/G]GGAGGCAGATCATCCGGGCTCTGTG
Phenotype
MIM: 605322
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
LOC105376777 PubMed Links

Gene Details

Gene
LOC105376777
Gene Name
uncharacterized LOC105376777
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
XM_011523512.2 300 Intron XP_011521814.1
Gene
WFDC1
Gene Name
WAP four-disulfide core domain 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001282466.1 300 Missense Mutation AGG,GGG R12G NP_001269395.1
NM_001282467.1 300 Missense Mutation AGG,GGG R12G NP_001269396.1
NM_021197.3 300 Missense Mutation AGG,GGG R12G NP_067020.2
XM_005256079.3 300 Missense Mutation AGG,GGG R12G XP_005256136.1

View Full Product Details