Product Details

SNP ID
rs11542345
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.17:28655313 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGGTTTCCAGAAAGAGGTGAAGTGA[A/G]GTGGTGACTATGGAGGTTTCGGCCA
Phenotype
MIM: 602934 MIM: 601333
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
SDF2 PubMed Links

Gene Details

Gene
SDF2
Gene Name
stromal cell derived factor 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_006923.3 618 Missense Mutation CTC,TTC L108F NP_008854.2
XM_011525106.2 618 Missense Mutation CTC,TTC L108F XP_011523408.1
Gene
SUPT6H
Gene Name
SPT6 homolog, histone chaperone
There are no transcripts associated with this gene.

View Full Product Details