Product Details

SNP ID
rs8095846
Assay Type
Functionally Tested
NCBI dbSNP Submissions
NA
Location
Chr.18:6943529 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AGAGCTAACTAGGAGAAACAGAAAA[C/T]AGTTTTGAAAAGTAACCTTTTAAAG
Phenotype
MIM: 150320
Polymorphism
C/T, Transition Substitution
Allele Nomenclature
Literature Links
LAMA1 PubMed Links

Gene Details

Gene
LAMA1
Gene Name
laminin subunit alpha 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_005559.3 Intron NP_005550.2
XM_011525655.2 Intron XP_011523957.1
XM_011525656.2 Intron XP_011523958.1
XM_011525657.1 Intron XP_011523959.1

View Full Product Details