Product Details

SNP ID
rs7245606
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:36538282 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
ATCCTCGCTGATAGGAATAAAATAT[A/T]AATTAAACCTACAATTTTTACCTAG
Phenotype
MIM: 613749
Polymorphism
A/T, Transversion substitution
Allele Nomenclature
Literature Links
ZNF260 PubMed Links

Gene Details

Gene
ZNF260
Gene Name
zinc finger protein 260
There are no transcripts associated with this gene.

Gene
ZNF529
Gene Name
zinc finger protein 529
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001145649.1 Intron NP_001139121.1
NM_001145650.1 Intron NP_001139122.1
NM_001321351.1 Intron NP_001308280.1
NM_020951.4 Intron NP_066002.3
XM_006723302.3 Intron XP_006723365.1
XM_006723304.2 Intron XP_006723367.1
XM_011527164.2 Intron XP_011525466.1
XM_011527165.2 Intron XP_011525467.1
XM_011527166.2 Intron XP_011525468.1
XM_011527167.2 Intron XP_011525469.1
XM_011527169.2 Intron XP_011525471.1
XM_011527170.1 Intron XP_011525472.1
XM_017027039.1 Intron XP_016882528.1
XM_017027040.1 Intron XP_016882529.1
XM_017027041.1 Intron XP_016882530.1
XM_017027042.1 Intron XP_016882531.1
XM_017027043.1 Intron XP_016882532.1
XM_017027044.1 Intron XP_016882533.1
XM_017027045.1 Intron XP_016882534.1

View Full Product Details