Product Details

SNP ID
rs11552279
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.21:33903610 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CAACATATTTCTCGCCAATGCGCAC[A/C]ATCATTCCACCCAAGATTGACGGAT
Phenotype
MIM: 600828 MIM: 602442
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
ATP5O PubMed Links

Gene Details

Gene
ATP5O
Gene Name
ATP synthase, H+ transporting, mitochondrial F1 complex, O subunit
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001697.2 Intron NP_001688.1
Gene
ITSN1
Gene Name
intersectin 1
There are no transcripts associated with this gene.

View Full Product Details