Product Details
- SNP ID
-
hCV44037881
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.X:78113810 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- ATGGAGCCAAGTCGGTAGTCCTTAT[G/T]AGCCACCTAGGCCGGCCTGATGGTG
- Phenotype
-
MIM: 311800
- Polymorphism
- G/T, Transversion substitution
- Allele Nomenclature
-
- Literature Links
-
PGK1
PubMed Links
Gene Details
- Gene
- PGK1
- Gene Name
- phosphoglycerate kinase 1
There are no transcripts associated with this gene.
View Full Product Details