Product Details
- SNP ID
-
rs11541350
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.2:20447577 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- GTTCCCCGAGGTGTACGTGCCCACC[G/T]TCTTCGAGAACTATGTGGCCGACAT
- Phenotype
-
MIM: 165370
- Polymorphism
- G/T, Transversion substitution
- Allele Nomenclature
-
- Literature Links
-
RHOB
PubMed Links
Gene Details
- Gene
- RHOB
- Gene Name
- ras homolog family member B
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_004040.3 |
507 |
Missense Mutation |
GTC,TTC |
V38F |
NP_004031.1 |
View Full Product Details