Product Details

SNP ID
rs13107559
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.4:122167571 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TTCTCCTGTGCCGCCTCCTGCCCCT[A/G]CTGGCCATGGATGTTCTTCTCCTTC
Phenotype
MIM: 611565
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
KIAA1109 PubMed Links

Gene Details

Gene
KIAA1109
Gene Name
KIAA1109
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_015312.3 Intron NP_056127.2
XM_005263282.1 Intron XP_005263339.1
XM_005263287.1 Intron XP_005263344.1
XM_006714344.1 Intron XP_006714407.1
XM_011532319.1 Intron XP_011530621.1
XM_011532320.2 Intron XP_011530622.1
XM_011532321.2 Intron XP_011530623.1
XM_011532322.1 Intron XP_011530624.1
XM_011532323.1 Intron XP_011530625.1
XM_011532324.1 Intron XP_011530626.1
XM_011532325.1 Intron XP_011530627.1
XM_011532326.1 Intron XP_011530628.1
XM_011532327.1 Intron XP_011530629.1
XM_011532330.1 Intron XP_011530632.1
XM_017008695.1 Intron XP_016864184.1
XM_017008696.1 Intron XP_016864185.1
XM_017008697.1 Intron XP_016864186.1
XM_017008698.1 Intron XP_016864187.1
XM_017008699.1 Intron XP_016864188.1

View Full Product Details