Product Details

SNP ID
rs7565185
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.2:101274077 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TTTTTTTTTTTTAAGAGAAACACCC[A/G]TCAAATTTATTGCAGTTGTCCAGTC
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
CNOT11 PubMed Links

Gene Details

Gene
CNOT11
Gene Name
CCR4-NOT transcription complex subunit 11
There are no transcripts associated with this gene.

Gene
RNF149
Gene Name
ring finger protein 149
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_173647.3 Intron NP_775918.2
XM_005263920.2 Intron XP_005263977.1
XM_005263921.3 Intron XP_005263978.1
XM_011510990.2 Intron XP_011509292.1
Gene
SNORD89
Gene Name
small nucleolar RNA, C/D box 89
There are no transcripts associated with this gene.

View Full Product Details